Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calibration Weights, Class E2, Proof Line, Sartorius

Supplier: Sartorius
Sartorius Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.

  • 0 - 5 mg leaf weights: Aluminium
  • 10 - 500 mg leaf weights: Nickel silver
  • 1 g - 10 Kg Knob weights: Stainless steel

Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

E2 weights in plastic cases
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 0,030 Knob weight 1 SARBYCW312-00
1 0,030 Knob weight 1 SARBYCW312-AC-00
1 0,030 Knob weight 1 SARBYCW322-AC-00
2 0,040 Knob weight 1 SARBYCW322-00
2 0,040 Knob weight 1 SARBYCW352-AC-00
2 0,040 Knob weight 1 SARBYCW412-AC-00
5 0,050 Knob weight 1 SARBYCW352-00
5 0,050 Knob weight 1 SARBYCW422-AC-00
5 0,050 Knob weight 1 SARBYCW612-AC-00
10 0,060 Knob weight 1 SARBYCW412-00
10 0,060 Knob weight 1 SARBYCW622-AC-00
10 0,060 Knob weight 1 SARBYCW652-AC-00
20 0,080 Knob weight 1 SARBYCW422-00
20 0,080 Knob weight 1 SARBYCW712-AC-00
50 0,10 Knob weight 1 SARBYCW452-AC-00
100 0,16 Knob weight 1 SARBYCW512-AC-00
200 0,030 Knob weight 1 SARBYCW522-AC-00
500 0,80 Knob weight 1 SARBYCW552-AC-00
1 0,006 Wire weight 1 SARBYCW0121-00
2 0,006 Wire weight 1 SARBYCW0221-00
5 0,006 Wire weight 1 SARBYCW0521-00
10 0,008 Wire weight 1 SARBYCW1121-00
10 0,008 Wire weight 1 SARBYCW1221-00
50 0,012 Wire weight 1 SARBYCW1521-00
100 0,016 Wire weight 1 SARBYCW2121-00
200 0,020 Wire weight 1 SARBYCW2221-00
500 0,025 Wire weight 1 SARBYCW2521-00

E2 weights in wooden boxes
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 0,030 Knob weight 1 SARBYCW312-02
2 0,040 Knob weight 1 SARBYCW322-02
5 0,050 Knob weight 1 SARBYCW352-02
10 0,060 Knob weight 1 SARBYCW412-02
20 0,080 Knob weight 1 SARBYCW312-AC-02
20 0,080 Knob weight 1 SARBYCW422-02
50 0,10 Knob weight 1 SARBYCW452-AC-02
100 0,16 Knob weight 1 SARBYCW512-AC-02
200 0,030 Knob weight 1 SARBYCW322-AC-02
200 0,030 Knob weight 1 SARBYCW522-AC-02
500 0,80 Knob weight 1 SARBYCW552-AC-02
1 1,6 Knob weight 1 SARBYCW352-AC-02
1 1,6 Knob weight 1 SARBYCW412-AC-02
2 3,0 Knob weight 1 SARBYCW422-AC-02
5 8 Knob weight 1 SARBYCW612-AC-02
5 8 Knob weight 1 SARBYCW622-AC-02
10 16 Knob weight 1 SARBYCW652-AC-02
10 16 Knob weight 1 SARBYCW712-AC-02
1 0,006 Wire weight 1 SARBYCW0121-02
2 0,006 Wire weight 1 SARBYCW0221-02
50 0,012 Wire weight 1 SARBYCW1521-02
100 0,016 Wire weight 1 SARBYCW2121-02
200 0,020 Wire weight 1 SARBYCW2221-02
500 0,025 Wire weight 1 SARBYCW2521-02

E2 set of weights in plastic cases
Weight Tolerance Type Pk VWR Catalog Number Quantity
100 0,16 Knob weight 1 611-1704
200 0,30 Knob weight 1 611-1705
500 0,8 Knob weight 1 611-1706
1 1,6 Knob weight with glove 1 SARBYCW612-00
2 3,0 Knob weight with glove 1 SARBYCW622-00
5 8 Knob weight with glove 1 611-1709
10 16 Knob weight with glove 1 SARBYCW712-00

E2 set of weights in wooden boxes
Weight Tolerance Type Pk VWR Catalog Number Quantity
50 0,10 Knob weight 1 SARBYCW452-02
100 0,16 Knob weight 1 SARBYCW512-02
500 0,8 Knob weight 1 SARBYCW552-02
1 1,6 Knob weight with glove 1 SARBYCW612-02
2 3,0 Knob weight with glove 1 SARBYCW622-02
5 8 Knob weight with glove 1 SARBYCW652-02
10 16 Knob weight with glove 1 SARBYCW712-02
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty