Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calibration Weights, Class E2, Proof Line, Sartorius

Supplier: Sartorius
Sartorius Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.

  • 0 - 5 mg leaf weights: Aluminium
  • 10 - 500 mg leaf weights: Nickel silver
  • 1 g - 10 Kg Knob weights: Stainless steel

Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.



 

ORDER

Expand All / Collapse All

E2 weights in plastic cases
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 mg ±0.006 mg Leaf weight 1 SARBYCW0121-AC-00
2 mg ±0.006 mg Leaf weight 1 SARBYCW0221-AC-00
5 mg ±0.006 mg Leaf weight 1 SARBYCW0521-AC-00
10 mg ±0.008 mg Leaf weight 1 SARBYCW1121-AC-00
20 mg ±0.010 mg Leaf weight 1 SARBYCW1221-AC-00
50 mg ±0.012 mg Leaf weight 1 SARBYCW1521-AC-00
100 mg ±0.016 mg Leaf weight 1 SARBYCW2121-AC-00
200 mg ±0.020 mg Leaf weight 1 SARBYCW2221-AC-00
500 mg ±0.025 mg Leaf weight 1 SARBYCW2521-AC-00
1 g ±0.030 mg Knob weight 1 SARBYCW312-AC-00
2 g ±0.040 mg Knob weight 1 SARBYCW322-AC-00
5 g ±0.050 mg Knob weight 1 SARBYCW352-AC-00
10 g ±0.060 mg Knob weight 1 SARBYCW412-AC-00
20 g ±0.080 mg Knob weight 1 SARBYCW422-AC-00
50 g ±0.10 mg Knob weight 1 SARBYCW452-AC-00
100 g ±0.16 mg Knob weight 1 SARBYCW512-AC-00
200 g ±0.30 mg Knob weight 1 SARBYCW522-AC-00
500 g ±0.80 mg Knob weight 1 SARBYCW552-AC-00
1 kg ±1.6 mg Knob weight 1 SARBYCW612-AC-00
2 kg ±3.0 mg Knob weight 1 SARBYCW622-AC-00
5 kg ±8.0 mg Knob weight 1 SARBYCW652-AC-00
10 kg ±16 mg Knob weight 1 SARBYCW712-AC-00

E2 weights in wooden boxes
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 mg ±0.006 mg Leaf weight 1 SARBYCW0121-AC-02
2 mg ±0.006 mg Leaf weight 1 SARBYCW0221-AC-02
5 mg ±0.006 mg Leaf weight 1 SARBYCW0521-AC-02
10 mg ±0.008 mg Leaf weight 1 SARBYCW1121-AC-02
20 mg ±0.010 mg Leaf weight 1 SARBYCW1221-AC-02
50 mg ±0.012 mg Leaf weight 1 SARBYCW1521-AC-02
100 mg ±0.016 mg Leaf weight 1 SARBYCW2121-AC-02
200 mg ±0.020 mg Leaf weight 1 SARBYCW2221-AC-02
500 mg ±0.025 mg Leaf weight 1 SARBYCW2521-AC-02
1 g ±0.030 mg Knob weight 1 SARBYCW312-AC-02
2 g ±0.040 mg Knob weight 1 SARBYCW322-AC-02
5 g ±0.050 mg Knob weight 1 SARBYCW352-AC-02
10 g ±0.060 mg Knob weight 1 SARBYCW412-AC-02
20 g ±0.080 mg Knob weight 1 SARBYCW422-AC-02
50 g ±0.10 mg Knob weight 1 SARBYCW452-AC-02
100 g ±0.16 mg Knob weight 1 SARBYCW512-AC-02
200 g ±0.30 mg Knob weight 1 SARBYCW522-AC-02
500 g ±0.80 mg Knob weight 1 SARBYCW552-AC-02
1 kg ±1.6 mg Knob weight 1 SARBYCW612-AC-02
2 kg ±3.0 mg Knob weight 1 SARBYCW622-AC-02
5 kg ±8.0 mg Knob weight 1 SARBYCW652-AC-02
10 kg ±16 mg Knob weight 1 SARBYCW712-AC-02

E2 set of weights in plastic cases
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 mg ±0.006 mg Wire Weight 1 SARBYCW0121-00
2 mg ±0.006 mg Wire Weight 1 SARBYCW0221-00
5 mg ±0.006 mg Wire Weight 1 SARBYCW0521-00
10 mg ±0.008 mg Wire Weight 1 SARBYCW1121-00
20 mg ±0.010 mg Wire Weight 1 SARBYCW1221-00
50 mg ±0.012 mg Wire Weight 1 SARBYCW1521-00
100 mg ±0.016 mg Wire Weight 1 SARBYCW2121-00
200 mg ±0.020 mg Wire Weight 1 SARBYCW2221-00
500 mg ±0.025 mg Wire Weight 1 SARBYCW2521-00
1 g 0,030 ± mg Knob weight 1 SARBYCW312-00
2 g 0,040 ± mg Knob weight 1 SARBYCW322-00
5 g 0,050 ± mg Knob weight 1 SARBYCW352-00
10 g 0,060 ± mg Knob weight 1 SARBYCW412-00
20 g 0,080 ± mg Knob weight 1 SARBYCW422-00
100 g 0,16 ± mg Knob weight 1 611-1704
200 g 0,30 ± mg Knob weight 1 611-1705
500 g 0,8 ± mg Knob weight 1 611-1706
1 kg 1,6 ± mg Knob weight 1 SARBYCW612-00
2 kg 3,0 ± mg Knob weight 1 SARBYCW622-00
5 kg 8 ± mg Knob weight 1 611-1709
10 kg 16 ± mg Knob weight 1 SARBYCW712-00

E2 set of weights in wooden boxes
Weight Tolerance Type Pk VWR Catalog Number Quantity
1 mg ±0.006 mg Wire Weight 1 SARBYCW0121-02
2 mg ±0.006 mg Wire Weight 1 SARBYCW0221-02
50 mg ±0.012 mg Wire Weight 1 SARBYCW1521-02
100 mg ±0.016 mg Wire Weight 1 SARBYCW2121-02
200 mg ±0.020 mg Wire Weight 1 SARBYCW2221-02
500 mg ±0.025 mg Wire Weight 1 SARBYCW2521-02
1 g ±0.030 mg Knob weight 1 SARBYCW312-02
2 g ±0.040 mg Knob weight 1 SARBYCW322-02
5 g ±0.050 mg Knob weight 1 SARBYCW352-02
10 g ±0.060 mg Knob weight 1 SARBYCW412-02
20 g ±0.080 mg Knob weight 1 SARBYCW422-02
50 g ±0.10 mg Knob weight 1 SARBYCW452-02
100 g ±0.16 mg Knob weight 1 SARBYCW512-02
500 g ±0.80 mg Knob weight 1 SARBYCW552-02
1 kg ±1.6 mg Knob weight 1 SARBYCW612-02
2 kg ±3.0 mg Knob weight 1 SARBYCW622-02
5 kg ±8.0 mg Knob weight 1 SARBYCW652-02
10 kg ±16 mg Knob weight 1 SARBYCW712-02
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories for Calibration Weights, E1, E2, F1, F2, Sartorius
Description Pk VWR Catalog Number Quantity
Brush, extra large, 250 mm 1 SARBYAW14
Brush, large, 150 mm 1 SARBYAW13
Forceps with silicone-coated
tips, 230 mm, for 1 g - 1 kg
1 SARBYAW33
Forceps with silicone-coated tips, 115 mm, for 1 mg - 5 g 1 SARBYAW31
Forceps with silicone-coated tips, 160 mm, for 1 g - 200 g 1 SARBYAW32
Glass bell jar with support plate, for 100 g - 1 kg (2 kg) 1 SARBYAW02
Glass bell jar with support plate, for 10 kg 1 SARBYAW04
Glass bell jar with support plate, for 1 mg - 50 g (100 g or 200 g) 1 SARBYAW01
Glass bell jar with support plate, for 20 kg 1 SARBYAW05
Glass bell jar with support plate, for 2 kg - 5 kg 1 SARBYAW03
Handle, for lifting 10 kg weights 1 SARBYAW51
Handle, for lifting 20 kg weights 1 SARBYAW52
Handle, for lifting 50 kg weights 1 SARBYAW53
Handle, for lifting 5 kg weights 1 SARBYAW50
Pair of gloves, cotton 1 PAIR SARBYAW21
Pair of gloves, fine leather 1 PAIR SARBYAW22
Weight fork, for 1 kg 1 SARBYAW42
Weight fork, for 2 kg 1 SARBYAW43
Weight fork, for 500 g 1 SARBYAW41
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty