Would you like to log in again?
Beakers, low form, Corning®
Supplier: Corning Life Sciences
Corning®
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
With printed with blue graduations.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Capacity Height O.D. Graduation interval Pk VWR Catalog Number Quantity
5000 247 181 mm 500 ml 4 CORN1000P-5L
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty