Barbituric acid ≥98%, J.T.Baker®
Supplier: Avantor
|
Warning
|
B654-05EA
17987.73
INR
B654-05
Barbituric acid ≥98%, J.T.Baker®
Barbituric acid
Formula:
C₄H₄N₂O₃ MW: 128,09 g/mol Boiling Pt: 441 °C (1013 hPa) Melting Pt: 250…252 °C Density: 0,95 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00006666 CAS Number: 67-52-7 EINECS: 200-658-0 Merck Index: 13,00966 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Drying at 105°C | ≤2 % |
Solubility (4% in H₂O) | Passes Test |
Assay (C₄H₄N₂O₃) (by acid-base titration) | ≥98 % |
Appearance | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...