Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stereo zoom microscopes, VisiScope® 350

Supplier: VWR Collection
VWR® VisiScope®
The VisiScope 350 can be used for observations in advanced teaching, laboratory and industrial applications.

  • Greenough optical system
  • High LED brightness with intensity control using a knob on the base
  • Dual-halogen illumination and intensity control with two knobs on the right side of the base and adjustable reflexion angle

Ordering information: A cleaning and maintenance set is available.




 

ORDER

Expand All / Collapse All
Description Type Model Pk VWR Catalog Number Quantity
Stereo zoom microscope with overhanging arm Binocular SZB350 OH 1 630-1583
Stereo zoom microscope Binocular SZB350 1 630-1578
Stereo zoom microscope with pillar Binocular SZBL350 1 630-1942
Stereo zoom microscope with overhanging arm Trinocular SZT350 OH 1 630-1584
Stereo zoom microscope Trinocular SZT350 1 630-1580
Stereo zoom microscope with pillar Trinocular SZTL350 1 630-1943

Eyepieces
Description Magnification Pk VWR Catalog Number Quantity
Widefield eyepieces 10× (20 mm) 2 630-1779
Eyepieces 15× (15 mm) 2 630-1780
Widefield eyepieces 20× (10 mm) 2 630-1781
Widefield micrometric eyepiece 10× (20 mm) 1 630-1782

Lenses
Description Magnification Working distance Pk VWR Catalog Number Quantity
Lens for stereo microscopes 0,5× 165 mm 1 630-1783
Lens for stereo microscopes 0,75× 117 mm 1 630-1784
Lens for stereo microscopes 1,5× 47 mm 1 630-1785
Lens for stereo microscopes 26 mm 1 630-1786
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model SZB350 OH SZT350 OH SZB350 SZT350 SZTL350 SZBL350
Eyepieces WF 10×/20 mm
Head 45° inclined, 360° rotating
Zoom lens 0,7× - 4,5×
Working distance 100 mm
Illumination Halogen LED
Stage Clips and frosted disc
Focusing system Pair of knobs on each side of the stand

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Halogen bulb with dichroic mirror 12 V/20 W 1 630-1798
Darkfield condenser 1 630-1788
White and black plate (Ø 95 mm) 1 630-1795
Glass stage (Ø 95 mm) 1 630-1796
Halogen bulb 12 V/15 W 1 630-1797
Polarising set with filters and rotating stage 1 630-1787
Sample clip 1 630-1789
Dust cover (L) for VisiScope® IT400 and stereo with overhanging stand (L×H) 700×550 mm 1 630-2824
Eyecups (Ø = 95 mm) 1 630-1794
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty