AMRE0885
Supplier: VWR Chemicals
0885-1GEA 20065 INR
VWRC0885-1G VWRC0885-100MG VWRC0885-500MG
AMRE0885
Chromogenic substrate for the detection of alkaline phosphatase. Often used with INT or NBT to amplify the sensitivity of staining.