Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetracycline hydrochloride, ultrapure

Supplier: VWR Chemicals
Warning

Blocks the binding of tRNA to the 30S subunit. Recommended working concentration: 15 μg/ml.


Formula: C₂₂H₂₅ClN₂O₈
MW: 480,9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8
UN: 0000
Specification Test Results
4-Ephianhydrotetracycline 2.0 %
Crystallinity PASS
Expiration Date REPORT
Heavy Metals 0.005 %
Identification PASS
Loss on Drying 2.0 %
pH (1%, Water) @25 °C 1.8 - 2.8
Potency (Anhydrous) 900 mcg/mg
Potency (As Is) REPORT
Specific Rotation -255 to -240 °

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 25 g 0422-EU-25G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty