Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kanamycin sulphate, ultrapure

Supplier: VWR Chemicals
Danger
Synonyms: Kanamycin AKanamycin Monosulphate

Binds to the 70S subunit of bacterial ribosome. Recommended working concentration: 30 μg/ml.


Formula: C₁₈H₃₈N₄O₁₅S
MW: 582,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
UN: 0000
Specification Test Results
Chromatographic Purity PASS
Crystallinity PASS
Expiration Date REPORT
Identification PASS
Loss on Drying 4.0 %
pH (1%, Water) @25°C 6.5 - 8.5
Potency (Anhydrous) 750 mcg/mg
Potency (As is) REPORT
Residue on Ignition 1.0 %

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle for solids 10 g 0408-EU-10G
Plastic bottle for solids 25 g 0408-EU-25G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty