Streptomycin sulphate, Ultra Pure Grade

Warning

0382-EU-50GEA 0 INR
0382-EU-50G
Streptomycin sulphate, Ultra Pure Grade
Streptomycin sulphate

30 μg/ml, binds to the 30S subunit of bacterial ribosome.

Formula: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
MW: 1457,4 g/mol
Melting Pt: 250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037023
CAS Number: 3810-74-0
EINECS: 223-286-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08983

Order Now

Specification Test Results

Loss on Drying (USP) <= 5 %
pH (20%, Water) @25°C 4.5 - 7
Potency (Dry Basis) 650 mcg/mg - 850 mcg/mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR