Neomycin sulphate, Ultra Pure Grade
Supplier: VWR Chemicals
Synonyms:
Fradiomycin sulphate
, Neomycin
, Neomycin sulphate salt
, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate
0558-EU-25GEA
6094
INR
0558-EU-25G
Neomycin sulphate, Ultra Pure Grade
Neomycin sulphate
Antibiotic. Causes miscoding during protein synthesis. Recommended working concentration: 50 μg/ml.
Formula:
C₂₃H₄₆N₆O₁₃·3H₂SO₄ MW: 908,87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150827 CAS Number: 1405-10-3 EINECS: 215-773-1 Merck Index: 12,06542 |
Specification Test Results
Expiration Date | REPORT |
Identification | PASS |
Loss on Drying (%) | ≤ 8.0 |
pH (3.3%, Water) @ 25 °C | 5.0 – 7.5 |
Potency (Anhydrous) (mcg/mg) | ≥ 600 |
Potency (as is) (mcg/mg) | REPORT |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...