Specimen bags, biohazard, Whirl-Pak® Trans-Pak™
Supplier: Nasco
NASCB01253WAEA 0 INR
NASCB01253WA
Specimen bags, biohazard, Whirl-Pak® Trans-Pak™
This is unbreakable PE bag has a unique outside pouch to separate important patient paperwork from the inside of the bag and the specimen. This unique specimen security system has a positive closure that is guaranteed leakproof when properly closed.
  • Easy to handle
  • Non sterile
  • Minimum storage and carrying space
  • Positive closure with 'puncture proof' protects bags against piercing
Packaging: Supplied in box of 500 bags.