Thymidine (2'-deoxythymidine), Ultrapure

Danger

Synonyms: dT1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dioneThymine deoxyriboside

0481-25GEA 21255 INR
0481-25G
Thymidine (2'-deoxythymidine), Ultrapure
Thymidine (2'-Deoxythymidine)
Formula: C₁₀H₁₄N₂O₅
MW: 242,23 g/mol
Melting Pt: 187−189 °C
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS Number: 50-89-5
EINECS: 200-070-4

Order Now

Specification Test Results

Em (267 nm, Phosphate buffer, pH 7.0) ≥9200

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR