Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Streptozocin, ultrapure

Supplier: VWR Chemicals
Danger

Selection agent, mutagenic agent, and diabetes inducer.


Formula: C₈H₁₅N₃O₇
MW: 265,2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006607
CAS Number: 18883-66-4
EINECS: 242-646-8
UN: 3077
ADR: 9,III
Specification Test Results
Carbon 35.9 % - 36.6 %
Heavy Metals < 20 ppm
Loss on Drying < 1 %
Melting Point (with decomposition) > 121°C
Nitrogen 15.6 % - 16.2 %
Residue on Ignition < 0.1 %
Solubility (1%, Water) Pass
Specific Rotation > 39°
Residue on ignition 0.1 %
Solubility (1 %, Water) PASS
Specific Rotation +39 °

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 1 g N407-1G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty