Centrifuge, refrigerated, floor-standing, Cryofuge® 6000i / 8500i series
Supplier: Thermo Scientific
HERA75007561 HERA75007520 HERA75007562 HERA75007566
Centrifuge, refrigerated, floor-standing, Cryofuge® 6000i / 8500i series
These large volume, floor standing, refrigerated centrifuges offer an optimised technology for lowspeed applications.
  • SEPACONTROL® operating panel is systematically arranged, clearly displaying all functional areas and allowing easy access to operating parameters
  • Robust, automated lid-lock mechanism protects operators and makes lid openings and closures effortless
  • Intuitive operator keypad allows you to recall protocols at the touch of a button; only five keys are required for completely programming all parameters
  • Easy user access: Front panel controls allow users to start, stop and open the lid of the centrifuge
  • Energy-saving design increases refrigeration efficiency and minimizes heat output, ensuring a more comfortable working environment
  • Heranet® software allows users to document centrifugation in blood banks
Ideal for use in blood banks, biotechnology and the pharmaceutical industry, where large volumes require high speeds and constant temperatures.
The use of sophisticated microelectronics systems and high strength materials for rotors and accessories guarantees outstanding reproducibility and highly-accurate centrifugation.
Delivery information: Supplied with wind-shielded rotor with lid.