Would you like to log in again?
Spatulas, micro
Supplier: Corning Life Sciences
Corning®
PS, white. These micro spatulas feature a V-shaped scoop (capacity < 0,5 ml) at one end and either a flat tapered or a flat rounded blade at the other end.
  • Free from RNase and DNase, non pyrogenic
  • Anti-static and sterile
  • Each micro spatula is individually packaged
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Length Pk VWR Catalog Number Quantity
Spatula with V-shaped scoop and flat tapered end 150 50 734-1529
Spatula with V-shaped scoop and flat rounded end 150 50 734-1530
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty