Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ampicillin sodium salt for tissue culture, γ-irradiated

Supplier: VWR Chemicals
Danger
Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

Aseptically prepared, gamma-irradiated. Tissue culture tested. 10 mg/ml solution. Working concentration: 50 μg/ml


Formula: C₁₆H₁₈N₃NaO₄S
MW: 371,39
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1
UN: 0000
Specification Test Results
Expiration Date REPORT
pH (1%, Water) @25 °C 8.0 - 10.0
Potency (Anhydrous) 845 - 988 mcg/mg
Solubility (Vial Contents, 20 ml Water) PASS
Sterile (gamma-irradiated) PASS
USP Grade Ampicillin Sodium PASS

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Vial 20 ml E477-20ML
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty