Would you like to log in again?
Robotic tips, Axygen®, for Qiagen®
Supplier: Corning Life Sciences
Corning® Axygen®
Transparent or black PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid level sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
ORDER
Expand All / Collapse All
BioRobot®
Tip style Volume Sterile Filter Tip colour Compatible head Packed Pk VWR Catalog Number Quantity
QTF-1100-CBK-R-S 1000 µl + + Black Biorobot® 16 racks, 96 each 1.536 AXYGQTF-1100-CBKRS
Corbett®
Tip style Volume Sterile Filter Tip colour Compatible head Packed Pk VWR Catalog Number Quantity
CTF-199-R-S 200 µl + + Transparent Corbett® 50 racks, 96 each 4.800 736-0522
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty