Metal-free, lightweight safety shoe made from synthetic textiles, soft padded tongue and collar plus an extremely flexible, dual-density PUR sole (slip, abrasion and cut-resistant, resistant to oil and petrol also withstands brief exposure to heat of approx. 120 °C).
- Climazone technology for improved comfort
- Breathable textile lining
- Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
- Anti-twist heel cap for better stability and protection from ankle twisting
- uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
- Virtually seamless design eliminates potential pressure points and blisters
- 100% metal-free xenova® toecap, non magnetic (200 J)
- Extremely flexible, lightweight PUR sole, which is abrasion, cut and slip resistant
- Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome, made from synthetic textiles.
Colour: White.
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S2, SRC