Would you like to log in again?
Colony count pen
Supplier: VWR Collection
Pen style electronic colony counter for rapid and accurate counts. Low energy consumption: Up to  1 000 000 counts before changing the battery. This electronic pen counter is ideal to ensure precise counts. The lightweight and ergonomic shape make heavy workload operation easy. The colony counting pen can be reset at any time by pressing the white button or turning off the pen. Once the counting is complete, the pen can be used to record the counts. A low cost way to prevent missing or double counting colonies.
  • 4-figure digital display
  • Audible alarm for end of count
  • Reset button
  • Light and ergonomic
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Colony count pen 1 710-0884
Refill for electronic colony counter pen 5 710-0902
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty