Homogeniser, HO 4/A
Supplier: Edmund Bühler
432-2111EA 0 INR
BUHL3301000
Homogeniser, HO 4/A
Homogeniser for mechanical crushing of samples (dry or in suspension) with wing knives at very high speed. Well suited for greasy/brittle, moist/fibrous as well as hard/brittle substances up to Mohs’ hardness of 5.
Delivery information: Basic device includes protection beaker with safety lock, 2 open-end wrenches for fastening the accessories.