Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

Wipe Dispensers

6  results were found
https://in.vwr.com/stibo/smallweb/std.lang.all/96/88/18709688.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Collection
Description:   Made from 6 mm white PVC.
Supplier:  VWR Collection
Description:   These dispensers, made from white lacquered metal, are suitable for use with wipe rolls.
Catalog Number: (115-2179)

Supplier:  KIMBERLY CLARK
Description:   This roll dispenser can be used with 250 mm paper towel rolls.
UOM:  1 * 1 items
Catalog Number: (115-1441)

Supplier:  Brand
Description:   Dispenser box, Bench top, Pursept® Wipes XL, For: Pursept® Wipes XL, Plastic
UOM:  1 * 1 items
Catalog Number: (115-1389)

Supplier:  FHP VILEDA SCS
Description:   Dispenser, Wall-mountable, For: Rolls, Plastic, White/clear
UOM:  1 * 4 items
Supplier:  SCA
Description:   W1: Tork Performance dispensers for large rolls are ergonomically designed dispensers with safe, reliable tear-off, easy load feature and extra...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 6  of 6