Would you like to log in again?
Bottles, wide neck, square, with screw cap
Supplier: Bürkle
Polycarbonate crystal-clear, with PP screw cap.
  • Autoclavable
  • Guaranteed leakproof
  • Moulded-in raised graduation
  • Shatterproof at room temperature
High-quality laboratory bottle made of polycarbonate for universal use. Easily operated screw thread, safe pouring,
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Capacity Thread Height W×D×H Neck I-Ø Pk VWR Catalog Number Quantity
1000 ml 37 mm 218 mm 95×95×218 mm 28 mm 1 BURK0316-1000
Get a discount if you order 4 or more. Login in now for details.
2000 ml 53 mm 275 mm 116×116×275 mm 39 mm 1 BURK0316-2000
Get a discount if you order 6 or more. Login in now for details.
250 ml 37 mm 145 mm 61×61×145 mm 28 mm 1 BURK0316-0250
Get a discount if you order 6 or more. Login in now for details.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty