Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus(V) oxide, Puratronic®, 99.9925% (metals basis)

Supplier: Alfa Aesar
Danger

Purity: 99.9925%
CAS No.: 1314-56-3
Molecular Formula: P2O5
Formula Weight: 141.94
Physical Form: Powder
Density: 2.39
Melting Point: 340°
Boiling Point: 360° subl.
Merck Reference: 14,7355
UN No.: 1807
MDL No.: MFCD00011440


Formula: P₂O₅
MW: 141,94 g/mol
Melting Pt: 562 °C
Density: 2,253 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011440
CAS Number: 1314-56-3
EINECS: 215-236-1
UN: 1807
ADR: 8,II
Merck Index: 13,07441

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
- 5 10827.59
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Density 2.39
Sensitivity Moisture Sensitive
Melting point 340°
Formula weight 141.94
CAS No. 1314-56-3
Purity 99.9925%
Physical form Powder
Odour/scent Weak
Boiling point 360° subl.
Molecular formula P2O5
MDL No. MFCD00011440
UN No. 1807
Merck reference 14,7355
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty