You Searched For: Barium chloride


7  results were found

SearchResultCount:"7"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (87783.290)
Supplier: VWR Chemicals
Description: Barium chloride 0.25 mol/l (0.5 N) in aqueous solution Reag. Ph. Eur. 1009301
UOM: 1 * 1 L

Catalog Number: (87784.290)
Supplier: VWR Chemicals
Description: Barium chloride 0.15 mol/l (0.3 N) in aqueous solution Reag. Ph. Eur. 1009302
UOM: 1 * 1 L

Supplier: VWR Chemicals
Description: Ready to use test/reagent solutions (TS and RS).
Catalog Number: (164735P)
Supplier: VWR Chemicals
Description: Barium chloride 10% in aqueous solution
UOM: 1 * 1 L

Catalog Number: (0980-01)
Supplier: Avantor
Description: Barium chloride, anhydrous ≥98.0%, powder, purified, J.T.Baker®
UOM: 1 * 500 g

Catalog Number: (30594.298)
Supplier: VWR Chemicals
Description: Barium chloride 1 g/l, VOLUSOL® for hydrometry
UOM: 1 * 1 L

MSDS


Catalog Number: (5907-01)
Supplier: Avantor
Description: Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®
UOM: 1 * 500 mL

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7 of 7
no targeter for Bottom