Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Sulphuric acid (≥ 95%)"

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Sulphuric acid 95% ARISTAR® for trace analysis
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   Sulphuric acid (≥ 95%) 98% for nitrogen analysis
Supplier:  VWR Chemicals
Description:   Sulphuric acid 95% AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Description:   Sulphuric acid 98% AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Description:   SULFURIC ACID 95.0 - 98.0% ACS GRADE 1 * 100 ml
Supplier:  VWR Chemicals
Description:   Sulphuric acid 95% GPR RECTAPUR®
Catalog Number: (20685.330)
Supplier:  VWR Chemicals
Description:   Sulphuric acid 95% TECHNICAL
UOM:  1 * 2,5 l
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7