You Searched For: Periodic acid


4  results were found

SearchResultCount:"4"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Chemicals
Description: Periodic acid ≥97%, GPR RECTAPUR®
Catalog Number: (294604D)
Supplier: VWR Chemicals
Description: Periodic acid 50%, GPR RECTAPUR®
UOM: 1 * 100 mL

Catalog Number: (87887.180)
Supplier: VWR Chemicals
Description: Periodic acid 0.002 mol/l in acetic acid Reag. Ph. Eur. 1063000
UOM: 1 * 100 mL

Supplier: VWR Chemicals
Description: Periodic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 4 of 4
no targeter for Bottom