Cesium chloride

Synonyms: Cesium Chloride

A1126.1000EA 0 INR
A1126.1000 A1126.0100
Cesium chloride
Cesium chloride
Formula: CsCl
MW: 168,36 g/mol
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 646 °C
Density: 3,7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR