Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Cesium chloride

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"7"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   A high purity reagent used to prepare density gradients for isolation of plasmid DNA, phage or cellular RNA.
Supplier:  VWR Chemicals
Description:   Cesium chloride ≥99.5%, AnalaR NORMAPUR® analytical reagent
UOM:  1 * 100 g
Supplier:  VWR Chemicals
Description:   Cesium chloride ≥99.5%, Electran Molecular biology grade

Supplier:  Avantor Performance Materials
Description:   Cesium chloride, ultrapure Bioreagent
UOM:  1 * 100 g
Supplier:  VWR Chemicals
Description:   Cesium chloride ≥99.99%, Gen-Apex, high purity Molecular biology grade
UOM:  1 * 1 kg
Supplier:  PanReac AppliChem
Description:   Cesium chloride 99.9 % BioChemica 1 * 500 g
Catalog Number: (A1098.1000)

Supplier:  PanReac AppliChem
Description:   Cesium chloride 99.999 % Molecular biology grade 1 * 1 kg
UOM:  1 * 1 kg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7