Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylcyclohexane ≥99%

Supplier: Alfa Aesar
Danger

Purity: 99%
CAS No.: 108-87-2
Molecular Formula: C7H14
Formula Weight: 98.19
Flash Point: -4°(-25°F)
Refractive Index: 1.4230
Melting Point: -126°
Boiling Point: 101°
Beilstein Reference: 505972
Merck Reference: 14,6047
UN No.: 2296
MDL No.: MFCD00001497


Formula: C₆H₁₁CH₃
MW: 98,19 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: -126 °C
Density: 0,7693 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://in.vwr.com/stibo/web/std.lang.all/34/11/773411.jpg

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 500 ml A16057.AP
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point -126°
Formula weight 98.19
CAS No. 108-87-2
Purity 99%
Refractive index 1.4230
Flash point -4°(-25°F)
Boiling point 101°
Molecular formula C7H14
MDL No. MFCD00001497
UN No. 2296
Merck reference 14,6047
Beilstein reference 505972
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty