Adenine sulphate dihydrate ≥99%, high purity
Synonyms:
9H-Purin-6-amine hemisulphate dihydrate
, 6-Aminopurine sulphate dihydrate
, Adenine hemisulphate dihydrate
, 7H-Purin-6-amine hemisulphate dihydrate
0607-100GEA
12525.75
INR
0607-100G
Adenine sulphate dihydrate ≥99%, high purity
Adenine sulphate dihydrate
Component of YAC selection media.
Formula:
(C₅H₅N₅)₂·H₂SO₄·2H₂O MW: 404,37 g/mol Melting Pt: 205…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213655 CAS Number: 6509-19-9 EINECS: 206-286-5 |
Specification Test Results
Em (262 nm, 0.01 N HCl) | >13000 |
Purity | ≥99% |
Solubility (0.05%, water) | Pass |
Water (KF) | 6 – 10% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...