Would you like to log in again?
Bioassay dish, treated
Supplier: Corning Life Sciences
Optically clear PS. Treated for optimal cell attachment.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Recommended working volume Growth area W×D×H Pk VWR Catalog Number Quantity
Dish, square, TC treated 100 - 150 ml 500 cm² 245×245×25 mm 16 734-1727
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty