Would you like to log in again?
Refrigerators for chemicals
Supplier: Julabo
Refrigerators for chemicals are designed for storing and cooling hazardous substances. The spark free interior prevents damage caused by spilled or evaporating chemicals.
  • Self-protecting control circuit
  • Digital temperature display (LED)
  • Overload protection for cooling compressor with test button
  • Cut-off in case of disturbance with optical alarm signal
  • Working and safety sensors are protected against short circuits and disconnection
ORDER
Expand All / Collapse All
Type Pk VWR Catalog Number Quantity
Refrigerator for chemicals with spark free interior for storing and cooling of chemicals and hazardous substances, KRC 180 1 471-1127
Refrigerator for chemicals with spark free interior for storing and cooling of chemicals and hazardous substances, KRC 50 1 471-1126
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Model KRC50 KRC180
Capacity (l) 50 180
Temperature (°C) ‒2...+12
Display LED
Alarm visual
Doors 1
Specifications With 1 support rack With 4 support racks
W×D×H int. (mm) 415×310×390 520×400×700
W×D×H ext. (mm) 530×630×535 600×640×855
Weight (kg) 23 35
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty