Shell vials PP with PE plug
548-1856EA 21489 INR
VWRI548-1856
Shell vials PP with PE plug
These vials are suitable for use with instruments from the following manufacturers: Alcott, Gilson, Shimadzu and Waters® (Wisp 96 respectively 48-position carousel).
  • Vial/closure combination for non critical analyses
  • Star-shaped diaphragm enables easy PE plug penetration
  • Recommended for HPLC usage