Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dodecyl sulphate (SDS) ≥99% Molecular biology grade

Supplier: VWR Chemicals
Danger
Synonyms: Dodecyl sulphate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphate



Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288,38 g/mol
Melting Pt: 204-207 °C
Density: 1,1 g/cm³ (20 °C)
Flash Pt: > 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://in.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg
Specification Test Results
Assay Min. 99.00 %
Assay (C10 compounds) Max. 1.0 %
Assay (C12 compounds) Min. 98.0 %
Assay (C14 compounds) Max. 1 %
DNases Not detected
RNases Not detected
Proteases Not detected
Absorbance (260 nm) (0.1 mol/l) Max. 0.050
Absorbance (280 nm) (0.1 mol/l) Max. 0.050
Heavy metals (as Pb) Max. 0.0005 %
pH (10 %) 6.00 - 7.50
Solubility in ethanol Passes test
Water Max. 1.0 %
Cl (Chloride) Max. 0.005 %
PO4 (Phosphate) Max. 0.0001 %
Pb (Lead) Max. 0.0001 %

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 50 g 444462R
Glass bottle for solids 500 g 444464T
Get a discount if you order 6 or more. Login in now for details.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty