Would you like to log in again?
Mini horizontal electrophoresis system, Owl B1A EasyCast™
Supplier: Thermo Scientific
The Owl B1A EasyCast™ mini gel system is designed to provide flat, even banding patterns and consistent results with minimum effort. This system is ideal for determination of molecular weight ranges of PCR fragments or larger sized DNA molecules. The all-in-one design allows the option of casting and running a gel in the buffer chamber, eliminating the need for additional equipment.
  • Two comb slots on the UV transparent gel tray double sample capacity by doubling the number of sample wells
  • Heavy duty, one-piece combs eliminate comb assembly
  • UVT gel trays are silk screened with a fluorescent ruler for easy measurement of bands
Ordering information: Each system includes buffer chamber, SuperSafe™ lid with attached power leads, gasketed UVT gel tray and two combs (6- and 10-well, double-sided, 1,0/1,5 mm thick)
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Owl B1A EasyCast complete system, with buffer exchange port 1 730-0183
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Gel size W×L (mm) 70×80
Sample capacity 5 to 24 samples in 15 minutes at 150 volts
Buffer volume (ml) 400
Unit W×L×H (mm) 95×160×80
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Replacement gaskets, pair 2 730-0185
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty