Trichloroacetic acid (TCA)
|
Danger
|
Synonyms:
TCA
A1431.1000EA
0
INR
A1431.1000
Trichloroacetic acid (TCA)
Trichloroacetic acid (TCA)
Formula:
Cl₃CCOOH MW: 163,39 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: 54…56 °C Density: 1,63 g/cm³ (20 °C) Flash Pt: 113 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004177 CAS Number: 76-03-9 EINECS: 200-927-2 Merck Index: 12,09756 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...