3-Aminopyridine-4-carbonitrile 97%

Danger

H32550.MDEA 0 INR
H32550.MD H32550.03
3-Aminopyridine-4-carbonitrile 97%
3-Aminopyridine-4-carbonitrile
Formula: C₆H₅N₃
MW: 119,13 g/mol
Melting Pt: 135…138 °C
Storage Temperature: Ambient
MDL Number: MFCD04035596
CAS Number: 78790-79-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR