Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2,6-difluoro-3-nitrobenzene ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 3847-58-3
Molecular Formula: C6H2ClF2NO2
Formula Weight: 193.54
Melting Point: 45-49°
UN No.: 2923
MDL No.: MFCD00042201


Formula: C₆H₂ClF₂NO₂
MW: 193,54 g/mol
CAS Number: 3847-58-3
EINECS: 411-980-8
UN: 2923
ADR: 8,III


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g H33806.03
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 45-49°
Formula weight 193.54
CAS No. 3847-58-3
Purity 97%
Molecular formula C6H2ClF2NO2
UN No. 2923
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty