Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Benzyloxyphenyl)acetic acid ≥99%

Supplier: Alfa Aesar



Formula: (C₆H₅CH₂OC₆H₄)CH₂COOH
MW: 242,27 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00017540
CAS Number: 6547-53-1
EINECS: 229-463-9

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
- 25 g H50902.14
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 119-121°C
Formula weight 242.28
CAS No. 6547-53-1
Purity 99%
Storage temperature 2°C to 8°C
Molecular formula C15H14O3
MDL No. MFCD00017540
Physical form Crystalline powder
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty