Acetone-[D₆] (99.9% D)

Danger

Synonyms: Propanone-D6DMK-D6HexadeuteroacetoneAcetone-d6Dimethyl ketone-D6Acetone-[²H₆]

16797.50EA 0 INR
16797.09 16797.50
Acetone-[D₆] (99.9% D)
Acetone-[D6]

NMR solvent

Formula: D₃CC(O)CD₃
MW: 64,03 g/mol
Boiling Pt: 56 °C
Melting Pt: –94 °C
Density: 0,872
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044635
CAS Number: 666-52-4
EINECS: 211-563-9
UN: 1090
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR