VWR® Tubular Knit Mop Heads
Supplier: Avantor
129-0489EA 0 INR
129-0489 129-0487 129-0488
VWR® Tubular Knit Mop Heads
Mops
Tubular-knit, 100% polyester string mop heads feature excellent sorbency.
Appropriate for ISO Class 4 (FED-STD-209E Class 10/M2.5) conditions. Mop heads are double clear poly-bagged for cleanroom use.