Would you like to log in again?
Hygiene mask, VWR® Critical Cover® with ShieldMate® Face Shield
Supplier: VWR Collection
VWR® Critical Cover® ShieldMate®
Face splash protection and inhalation security in one design.
  • ShieldMate combination facemask and eye shield features a low glare, anti-fog treated, optically clear shield ultrasonically welded to the mask
  • An anti-fog foam strip on the mask eliminates eyewear fogging
  • Tie mask with Magic Arch® keeps the mask away from the nose and mouth creating a larger breathing chamber for a cool and comfortable performance
  • Comfortable ties and a full-length aluminum nosepiece comfortably secure the ShieldMate mask to the face
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Type Colour Pk VWR Catalog Number Quantity
With headband and face shield Blue 100 111-0621
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty