β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) trihydrate ≥97%, Reagent Grade

Supplier: VWR Chemicals

0455-5GEA 12326 INR
0455-5G
β-Nicotinamide adenine dinucleotide, oxidised form (NAD, oxidised form) trihydrate ≥97%, Reagent Grade
β-Nicotinamide adenine dinucleotide (NAD, oxidized form) hydrate
Formula: C₂₁H₂₇N₇O₁₄P₂·H₂O
Melting Pt: 140 to 142 °C
Storage Temperature: Freezer
MDL Number: MFCD00036253
CAS Number: 53-84-9
EINECS: 200-184-4
Merck Index: 12,06429

Order Now

Specification Test Results

A250/A260 0.78 − 0.88
A280/A260 0.17 − 0.27
Lamda max (pH 7.0) 259 − 260 nm
Moisture (by Karl Fischer) <4.0%
Purity (Anhydrous) 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR