Analogue stopwatch
609-0130EA 8616 INR
VWRI609-0130
Analogue stopwatch
Crown stopper with frame ring.
  • Copper chromium plated casing
  • ¹/₁₀₀ second timing
  • Counts up to 15 minutes
Delivery information: Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.