Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Multi-channel pipettes, mechanical, variable volume, Transferpette® S -8/-12

Supplier: Brand
Transferpette®
The Transferpette® S -8/-12 multi channel pipettes are the perfect manual pipettes for the most demanding applications in the lab and include all features requested by Life Science users working in the 96-well and 384-well microplate format.

  • Ergonomic finger rest for a relaxed grip accommodates right- and left-handed users; central pipetting button and separate tip ejector; one-handed operation without changing the hand position
  • 4-digit volume display for highest precision
  • Easy calibration technique providing calibration adjustments without tools, and a clear indication when the pipettes have been adjusted from factory settings
  • Individually replaceable nose cones and seals to simplify service for long instrument life; corrosion resistant pistons
  • Colour-coded for easy tip selection
  • Fully autoclavable

Compatible with the most common tips. The use of new innovative materials make the Transferpette® S -8/-12 a lightweight yet rugged multi channel pipette with a high degree of accuracy and precision at the same time.

CE-marked according to IVD-Directive 98/79 EC

* Filter tips

Ordering information: Transferpette® S -8 or Transferpette® S -12 supplied with DE-M marking, performance certificate, 2 x TipBox (filled with pipette tips from BRAND), 1 shelf/rack mount, 1 reagent reservoir, 1 mounting tool, silicone oil and 1 set of sealing rings made of FKM.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

8-Channel
Capacity Division Accuracy Imprecision For pipette tips Pk VWR Catalog Number Quantity
0,5 - 10 µl 0,01 µl ±1,6 % ≤1,0 % 0,1-20 / 0,5-20 / 1-50 / 0,1-1* / 0,5-10* / 1-20* µl 1 613-3700
10 - 100 µl 0,2 µl ±0,8 % ≤0,3 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3702
20 - 200 µl 0,2 µl ±0,8 % ≤0,3 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3703
30 - 300 µl 0,5 µl ±0,6 % ≤0,3 % 2-200 / 5-300 / 5-100* / 5-200* µl 1 613-3704
5 - 50 µl 0,1 µl ±0,8 % ≤0,4 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3701

12-Channel
Capacity Division Accuracy Imprecision For pipette tips Pk VWR Catalog Number Quantity
0,5 - 10 µl 0,01 µl ±1,6 % ≤1,0 % 0,1-20 / 0,5-20 / 1-50 / 0,1-1* / 0,5-10* / 1-20* µl 1 613-3705
10 - 100 µl 0,2 µl ±0,8 % ≤0,3 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3707
20 - 200 µl 0,2 µl ±0,8 % ≤0,3 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3708
30 - 300 µl 0,5 µl ±0,6 % ≤0,3 % 2-200 / 5-300 / 5-100* / 5-200* µl 1 613-3709
5 - 50 µl 0,1 µl ±0,8 % ≤0,4 % 2-200 / 5-300 / 2-20* / 5-100* / 5-200* µl 1 613-3706
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Individual stand for Transferpette® electronic / Transferpette® S -8/-12 single instrument 1 613-1682
Bench top rack for 6 Transferpette® S and Transferpette® S-8/-12 pipettes 1 613-3475
Shelf/rack mount for Transferpette® S single instrument 1 613-3476
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty