Monocular microscopes, MH/ML series
630-0783EA 0 INR
VWRI630-0783 VWRI630-0784 VWRI630-0781 VWRI630-0782 VWRI630-0758 VWRI630-0756 VWRI630-0757 VWRI630-0755
Monocular microscopes, MH/ML series
These microscopes are designed for use in schools and educational establishments.
  • Anti-theft screw on the eyepiece
  • Possibility to add a fourth 100× objective
  • Ergonomic and easy to use instrument
  • Retractable objectives help prevent damage
Delivery information: Supplied with objectives, a protective dust cover and one fuse.