Analogue thermo-hygrometer clock
620-0921EA 4821 INR
VWRI620-0921
Analogue thermo-hygrometer clock
Thermometer-hygrometer analogue clock features plastic casing and white dial face.
  • Three functions in one
  • Contemporary design with bold markings
−30…+60 °C (±2,0 °C); 0 - 100% RH (±5,0% RH)
Delivery information: Supplied with hook for wall mounting. Requires AA battery (not included).