VWR® Advanced 700, Hotplates
Supplier: VWR
VWR®
444-2865EA 0 INR
444-2865 444-2863 444-2869 444-2814 444-2867
VWR® Advanced 700, Hotplates
Heaters Hotplates
VWR® 700 series advanced hotplates deliver accurate and repeatable results. Low profile, space saving design takes up less space in the lab. Spill resistant housing channels fluids away from internal components. Front panel features easy to use controls, which allow users to dial-in adjustments.
  • LED display and microprocessor control
  • Over-temperature protection
  • On-board memory recalls last set temperature
  • Ceramic or aluminium top plate
  • Hot warning when plate is in use
Integral support rod holder with locking knob accepts the optional support rod and clamp kit (444-2844). Microprocessor control regulates heating functions. Samples are heated to temperature quickly and accurately. Ceramic tops feature a chemically resistant, reflective white surface that's easy to clean. Durable aluminum tops provide a more even heating surface and will not crack or chip. The LED display, which is easy to read, shows set point temperature and the microprocessor control provides repeatable and accurate results every time. For additional safety, a hot symbol light is illuminated when the heat is turned on and remains on until the top plate cools down. Over-temperature protection ensures the unit will automatically shut down if it exceeds the set temperature by 10 °C. Built-in memory allows users to recall the last set temperature even when the unit has been turned off.
Delivery information: Each unit is supplied complete with 2 plugs: EU and UK.
Caution: * Below 100 °C ±2 °C Environmental and sample conditions permitting.