Electroporation system, ECM® 630
Supplier: BTX
BTXI45-0051
Electroporation system, ECM® 630
The ECM® 630 is a powerful exponential decay wave electroporation system designed for in vitro and in vivo electroporation applications, including mammalian and plant cell transfections, drug delivery and bacteria and yeast transformations.
  • Precision Pulse™ provides the user with full flexibility in choosing a wide range of time constants
  • Over 200 protocols are available, including the ability to reproduce protocols from electroporators from other manufacturers
Delivery information: Each complete system includes ECM® 630 generator with universal power supply, safety stand, cuvettes (10 each of 1 mm, 2 mm and 4 mm gap sizes), and cuvette rack.