Semi-dry blotter
Supplier: CBS Scientific
730-0073EA 0 INR
CBSCEBU-4000
Semi-dry blotter
These semi-dry blotters are available for gel dimensions up to 200×200 mm. They are designed for transfer of either Western, Northern, or Southern blots reliably and quickly.
  • Durable and corrosion free, stainless steel cathode and platinum-coated titanium anode create a uniform electric field producing excellent transfers onto nitrocellulose or nylon substrates
  • Upper electrode housing is separate from the base, allowing it to be secured at the desired height with side hand screws
  • Designed to permit several gels to be transferred simultaneously
  • Blotting is accomplished by layering transfer membrane and gel between buffer saturated blotting paper, minimising the amount of buffer required
  • To prevent drawing current away from the gel during a transfer, the uncovered portion of the cathode is shielded with a Mylar® cut-out, which ensures that all applied current passes directly through the gel